Learn More
TYSND1 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-05923APC
Product Code. 18662156
  • € 407.00 / 0.10mL
Description
TYSND1 Polyclonal antibody specifically detects TYSND1 in Human samples. It is validated for Western Blot