Learn More
DuPont™ Tyvek™ IsoClean™ Frock with Bound Neck
Product Code. 16488363 Shop All DuPont Products
Click to view available options
Size:
Small
Medium
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 30
Product Code. 16488363
Brand: DuPont™ D15535580
Only null left
Add to basket
Add to basket
Delivers an ideal balance of protection, durability and comfort.
  • Provides an inherent barrier to particles, microorganisms and non-hazardous water-based light liquid splash
  • Breathable and exceptionally low linting
TRUSTED_SUSTAINABILITY
Specifications
Product Type Controlled Environments Lab Coat
Size X-Large
Material Polyethylene
Color White
Closure Type Snaps