Learn More
DuPont™ Tyvek™ IsoClean™ Unhooded Coveralls
Click to view available options
Size:
Medium
2X-Large
3X-Large
Unit Size:
Pack of 25
Description
- Delivers an ideal balance of protection, durability and comfort.
- Made of high density polyethylene using a patented flash spinning process.
- Breathable and exceptionally low linting.
- Garments and accessories made of clean-processed and sterile Tyvek™ IsoClean™ are typically used in cleanrooms within the biotech, pharmaceutical, medical device manufacturing, food processing, cosmetics industry as well as in other critical or controlled environments.
- Clean-processed and sterilised by gamma-irradiation to SAL of 10-6 (ISO 11137-1)
- Full traceability on all sterilized apparel with certificates of sterility available
- Suitable for use in GMP class A/B (ISO Class 5) clean rooms
- This garment is not CE certified as chemical protective clothing
Specifications
Specifications
| Product Type | Cleanroom Coveralls |
| Size | 3X-Large |
| Material | HDPE |
| Color | White |
| Ankle Style | Elastic |
| Cuff Style | Elastic |
| Closure Type | Front Zipper |
| Seam Type | Bound |
| Sterility | Sterile |
| Protection Class | ISO Class 5 |
| Show More |