Learn More
TAC Trade And Converting Tyvek™ Overshoe with PE Sole
Description
- Polyethylene sole
- Sole thickness of 50 micron
- Latex free
Specifications
Specifications
| Size | One size |
| Latex Free | Yes |
| Color | White |
| Sterility | Sterile |
| Closure Type | Tie tape |
| Thickness | 50 micron sole |
| Includes | Elastic hem and cuffs |
| Packaging Quantity | 50 pair/BX |