Learn More
DuPont™ Sterile Tyvek™ IsoClean Overboots
Description
Tyvek™ IsoClean
Disposable Overboots
- Sterile overboot in white
- Fixation ties
- Tunnelled elastics at ankles and gripper slip-retardant full sole
- Composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort
- Permeable to both air and water vapour, yet repels water-based liquids and aerosols
- Offers an excellent barrier against fine particles and fibers (down to micron in size) and is ultra-low-linting
- Silicon non-added
- Garments and accessories made of Tyvek™ IsoClean are used in the biotech, pharmaceutical, medical device manufacturing and electronics industries, as well as in other critical or controlled environments
Disposable Overboots
- This accessory is not CE certified as chemical protective clothing
- Clean laundered and gamma-sterilised
- Suitable for use in cleanroom class ISO 5 and higher
- Sterility assurance level (SAL) of 10-6
- Bound seams: tightly sewn seam is covered with garment fabric to reinforce seam protection and reduce potential for liquid and particle penetration
- Full traceability on all sterilized apparel with certificates of sterility available
Specifications
Specifications
| Material | Tyvek |
| Color | White |
| Disposable | Single-use |
| Sole Type | Slip-resistant |
| Seam Type | Reinforced seam protection |
| Quantity | 100 |