Learn More
Mettler Toledo™ U-Electrode, Universal
Product Code. 10260445 Shop All Mettler Toledo Products
Click to view available options
:
Size: Large
Product Code. 10260445
missing translation for 'mfr': Mettler Toledo™ 11107764
Completely remove electrostatic charge from your samples and tare containers and avoid distortion of your weighing results. Mettler Toledo™ U-Electrode, Universal enables fast dosing processes and substantially reduces the risk of contamination.
  • Large electrode for discharging all kinds of samples and tare containers with dimensions of 15x25cm
  • Improves weighing accuracy
  • Enables fast dosing
  • Reduces contamination risk
  • Can be freely placed next to your balance
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Laboratory Balance
Electrical Requirements 100 to 240 V 50/60 Hz
For Use With (Application) Discharging electrostatic charge
Size Large