missing translation for 'onlineSavingsMsg'
Learn More
Learn More
u-Plasminogen Activator/Urokinase Antibody (U-16), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-05160MFV610
This item is not returnable.
View return policy
Description
u-Plasminogen Activator/Urokinase Monoclonal antibody specifically detects u-Plasminogen Activator/Urokinase in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
Specifications
| u-Plasminogen Activator/Urokinase | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| U-16 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen | |
| ATF, EC 3.4.21, EC 3.4.21.73, plasminogen activator, urokinase, uPA, u-PA, U-plasminogen activator, urinary, urokinase-type plasminogen activator | |
| Native human uPA | |
| 0.1 mL | |
| Cancer | |
| 5328 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction