missing translation for 'onlineSavingsMsg'
Learn More
Learn More
U2AF1L3 (U2AF1L4) Monoclonal Antibody (OTI8B1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies CF810435
This item is not returnable.
View return policy
Description
U2AF1L3 (U2AF1L4) Monoclonal antibody specifically detects U2AF1L3 (U2AF1L4) in Human samples. It is validated for Western BlotSpecifications
| U2AF1L3 (U2AF1L4) | |
| Monoclonal | |
| See Label | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q8WU68 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 199746 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI8B1 | |
| Unconjugated | |
| U2AF1L4 | |
| FLJ35525; MGC33901; Splicing factor U2AF 26 kDa subunit; U2 auxiliary factor 26; U2 small nuclear RNA auxiliary factor 1 like 4; U2 small nuclear RNA auxiliary factor 1-like 3; U2 small nuclear RNA auxiliary factor 1-like 4; U2 small nuclear RNA auxiliary factor 1-like protein 3; U2 small nuclear RNA auxiliary factor 1-like protein 4; U2(RNU2) small nuclear RNA auxiliary factor 1-like 3; U2(RNU2) small nuclear RNA auxiliary factor 1-like protein 3; U2AF1L3; U2AF1L3V1; U2AF1L4; U2AF1-like 4; U2AF1-like protein 3; U2AF1RS3; U2AF1-RS3; U2af26 | |
| Full length human recombinant protein of human U2AF1L4 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction