missing translation for 'onlineSavingsMsg'
Learn More

U2AF35 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 415.00 - € 624.00

Specifications

Antigen U2AF35
Dilution Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18623016
View Documents
Novus Biologicals
NBP2-46797-25ul
25 μL
€ 415.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18650986
View Documents
Novus Biologicals
NBP2-46797
0.1 mL
€ 624.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

U2AF35 Polyclonal antibody specifically detects U2AF35 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

U2AF35
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Core ESC Like Genes, Stem Cell Markers
PBS (pH 7.2), 40% Glycerol
FP793, human U2 snRNP auxiliary factor small subunit10U2AF35DKFZp313J1712, RN, RNU2AF1,35-KD subunit, U2 auxiliary factor 35 kDa subunit, U2 small nuclear RNA auxiliary factor 1, U2 small nuclear RNA auxillary factor 1, U2 snRNP auxiliary factor small subunit
This antibody was developed against Recombinant Protein corresponding to amino acids: CAVSDVEMQEHYDEFFEEVFTEMEEKYG
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Polyclonal
Purified
RUO
Human
Q01081
7307
IgG
Immunogen affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.