missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX U2OS OXGR1 Gq Cell Line

Product Code. 17314749
Encompass
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
This item is not returnable. View return policy

Product Code. 17314749

Brand: DiscoverX 951017C3

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The U2OS OXGR1 Gq Cell Line measures OXGR1 (GPCR) activation via an increase in intracellular calcium. This cell line ships as two vials of cryopreserved cells.

The OXGR1 Gq cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. α-ketoglutarate) based activation of OXGR1 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses OXGR1 , a naturally Gq coupled GPCR. Upon activation of OXGR1 , the resulting calcium activity can be fluorescently assayed using the Calcium NoWash PLUS Detection Kit. This stable modified cell line is provided as two vials of cryopreserved cells.

Specifications

Assay Entry Calcium Signaling
Cell Line U2OS
For Use With (Application) Cell Lines
Primary Agonist α-ketoglutarate
Quantity 2 Vials
Target Entry OXGR1
Accession Number NM_080818.3
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.