missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UAP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
€ 446.00 - € 447.00
Specifications
| Antigen | UAP1 |
|---|---|
| Concentration | 1 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles, store in dark |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17197193
![]() |
Invitrogen™
PA5109264 |
100 μL |
€ 446.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17107203
![]() |
Invitrogen™
PA5109265 |
100 μL |
€ 447.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Positive Control: SiHa, 293T, rat brain tissue, human liver tissue, K562.
Converts UDP and GlcNAc-1-P into UDP-GlcNAc, and UDP and GalNAc-1-P into UDP-GalNAc. Isoform AGX1 has 2 to 3 times higher activity towards GalNAc-1-P, while isoform AGX2 has 8 times more activity towards GlcNAc-1-P.Specifications
| UAP1 | |
| -20°C, Avoid Freeze/Thaw Cycles, store in dark | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| UAP1 | |
| AA420407; AA437972; AgX; AGX1; AGX-1; AGX2; AGX-2; antigen X; ESTM38; fb63d12; RGD1561967; similar to UDP-N-acetylhexosamine pyrophosphorylase (Antigen X) (AGX) (Sperm-associated antigen 2); SPAG2; sperm associated antigen 2; sperm-associated antigen 2; testis tissue sperm-binding protein Li 37a; UAP1; UDP-N-acetylgalactosamine pyrophosphorylase; UDP-N-acetylglucosamine diphosphorylase 1; UDP-N-acetylglucosamine pyrophosphorylase; UDP-N-acetylglucosamine pyrophosphorylase 1; UDP-N-acetylhexosamine pyrophosphorylase; UDP-N-acetylhexosamine pyrophosphorylase 1; UDP-N-acteylglucosamine pyrophosphorylase 1; wu:fb63d12; zgc:55280 | |
| UAP1 | |
| Primary | |
| Affinity Chromatography |
| 1 mg/mL | |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q16222 | |
| 6675 | |
| Synthetic peptide within Human UAP1 aa 392-441. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title