missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UAP1L1 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98049G
This item is not returnable.
View return policy
Description
UAP1L1 Polyclonal specifically detects UAP1L1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| UAP1L1 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| EC 2.7.7, EC 2.7.7.-, EC 2.7.7.23, EC 2.7.7.27, RP11-229P13.18, UDP-N-acetylhexosamine pyrophosphorylase-like protein 1, UDP-N-acteylglucosamine pyrophosphorylase 1-like 1 | |
| Produced in rabbits immunized with E. coli-derived Human UAP1L1 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 91373 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction