missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UAP56 Antibody (OTI2C5), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74727AF750
This item is not returnable.
View return policy
Description
UAP56 Monoclonal specifically detects UAP56 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| UAP56 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| DDX39B | |
| Full length recombinant protein of human UAP56 (NP_542165) produced in HEK293T cells. | |
| Proteases & Other Enzymes | |
| 7919 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI2C5 | |
| Western Blot, Immunohistochemistry | |
| 56 kDa U2AF65-associated protein, ATP-dependent RNA helicase p47, DEAD (Asp-Glu-Ala-Asp) box polypeptide 39B, DEAD box protein UAP56, EC 3.6.4.13, HLA-B associated transcript 1, HLA-B-associated transcript 1 protein, spliceosome RNA helicase BAT1, UAP56D6S81EBAT1nuclear RNA helicase (DEAD family) | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction