missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBA3 Antibody (S05-6C8), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 421.00 - € 645.00
Specifications
| Antigen | UBA3 |
|---|---|
| Clone | S05-6C8 |
| Concentration | 0.3 mg/mL |
| Dilution | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 10 μg/ml, Immunohistochemistry-Paraffin |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18658484
|
Novus Biologicals
NBP3-20008-50ul |
50 μg |
€ 421.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18686014
|
Novus Biologicals
NBP3-20008-100ul |
100 μg |
€ 645.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UBA3 Monoclonal antibody specifically detects UBA3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| UBA3 | |
| 0.3 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cell Cycle and Replication, Neuroscience | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 9039 | |
| IgG | |
| Affinity purified |
| S05-6C8 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 10 μg/ml, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DKFZp566J164, EC 6.3.2, EC 6.3.2.-, hUba3, MGC22384, NEDD8-activating enzyme E1 catalytic subunit, NEDD8-activating enzyme E1C, Nedd8-activating enzyme hUba3, UBA3, ubiquitin-activating enzyme E1 homolog, UBE1C, Ubiquitin-activating enzyme 3, Ubiquitin-activating enzyme E1C, ubiquitin-activating enzyme E1C (homologous to yeast UBA3), ubiquitin-like modifier activating enzyme 3, Ubiquitin-like modifier-activating enzyme 3, yeast) | |
| A synthetic peptide of human UBA3 (Uniprot # Q8TBC4) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title