missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBA52 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99383-50ul
This item is not returnable.
View return policy
Description
UBA52 Polyclonal antibody specifically detects UBA52 in Human samples. It is validated for Western Blot
Specifications
| UBA52 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| CEP52ubiquitin carboxyl extension protein 52, HUBCEP52, MGC126879, MGC126881, MGC57125, RPL40, UBCEP2, ubiquitin A-52 residue ribosomal protein fusion product 160S ribosomal protein L40, ubiquitin-52 amino acid fusion protein, ubiquitin-60S ribosomal protein L40, ubiquitin-CEP52 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human UBA52. | |
| 50 μg | |
| Apoptosis, Cell Biology | |
| 7311 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction