missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBA52 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 280.00 - € 556.50
Specifications
| Antigen | UBA52 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18251214
|
Novus Biologicals
NBP2-55293 |
100 μL |
€ 589.00 € 556.50 / 100µL Save € 32.50 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18697187
|
Novus Biologicals
NBP2-55293-25ul |
25 μL |
€ 280.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UBA52 Polyclonal specifically detects UBA52 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| UBA52 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| CEP52ubiquitin carboxyl extension protein 52, HUBCEP52, MGC126879, MGC126881, MGC57125, RPL40, UBCEP2, ubiquitin A-52 residue ribosomal protein fusion product 160S ribosomal protein L40, ubiquitin-52 amino acid fusion protein, ubiquitin-60S ribosomal protein L40, ubiquitin-CEP52 | |
| UBA52 | |
| IgG | |
| Affinity Purified |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 7311 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:RGGIIEPSLRQLAQKYNCDKMICRKCYARLHPRAVNCRKKKCGHTNNLRPKKKVK | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title