missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBA52 Antibody (OTI4F2), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74730FR
This item is not returnable.
View return policy
Description
UBA52 Monoclonal specifically detects UBA52 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| UBA52 | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| UBA52 | |
| Synthetic peptide corresponding to residues near C-terminus of human UBA52 (NP_003324). | |
| 0.1 mL | |
| 7311.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI4F2 | |
| Western Blot, Immunohistochemistry | |
| CEP52ubiquitin carboxyl extension protein 52, HUBCEP52, MGC126879, MGC126881, MGC57125, RPL40, UBCEP2, ubiquitin A-52 residue ribosomal protein fusion product 160S ribosomal protein L40, ubiquitin-52 amino acid fusion protein, ubiquitin-60S ribosomal protein L40, ubiquitin-CEP52 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction