missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBA52 Monoclonal Antibody (OTI1C6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805019S
This item is not returnable.
View return policy
Description
UBA52 Monoclonal antibody specifically detects UBA52 in Human samples. It is validated for Western BlotSpecifications
| UBA52 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P62987 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7311 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI1C6 | |
| Unconjugated | |
| UBA52 | |
| 60S ribosomal protein L40; CEP52; D8Ertd21e; Gm1863; HUB L40; HUBCEP52; L40; Large ribosomal subunit protein eL40; MGC127041; RPL40; RPS27A; Uba52; UBB; Ubc; Ubcep2; Ubiquitin; Ubiquitin A-52 residue ribosomal protein fusion product 1; ubiquitin and ribosomal protein L40; ubiquitin carboxyl extension protein 52; ubiquitin/60S ribosomal fusion protein; ubiquitin-52 amino acid fusion protein; ubiquitin-60S ribosomal protein L40; ubiquitin-CEP52; Unknown (protein for MGC:127041) | |
| Human recombinant protein fragment corresponding to amino acids 77-128 of human UBA52 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction