missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UBAC1 Monoclonal Antibody (OTI6E9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527282
This item is not returnable.
View return policy
Description
UBAC1 Monoclonal Antibody for Western Blot
UBAC1 belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein has two RRM domains that bind to RNAs.
Specifications
| UBAC1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9BSL1 | |
| UBAC1 | |
| Full length human recombinant protein of UBAC1 produced in 293T. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| OTI6E9 | |
| Unconjugated | |
| UBAC1 | |
| 1110033G07Rik; AA407978; E3 ubiquitin-protein ligase subunit KPC2; GBDR1; Gdbr1; glialblastoma cell differentiation-related protein 1; Kip1 ubiquitination-promoting complex protein 2; KPC2; putative glialblastoma cell differentiation-related; putative glialblastoma cell differentiation-related protein; RP11-432J22.3; testicular secretory protein Li 64; UBA domain containing 1; UBA domain-containing protein 1; Ubac1; Ubadc1; ubiquitin associated domain containing 1; ubiquitin-associated domain-containing protein 1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10422 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction