Learn More
UBAP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA554085
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: FHGTFSYLDD VPFKTGDKFK TPAKVGLPIG FSLPDCLQVV REVQYDFSLE KKTIEWAEEI KKIEEAEREA ECKIAEAEAK VNSKSGPEGD SKMSFSKTHS Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.
Originally named because of its sequence similarity to the Saccharomyces cerevisiae NHP2 (non-histone protein 2), this protein appears to be a highly conserved nuclear protein that is a component of the [U4/U6.U5] tri-snRNP. It binds to the 5' stem-loop of U4 snRNA. Two transcript variants encoding the same protein have been found for this gene.Specifications
UBAP1 | |
Polyclonal | |
Unconjugated | |
UBAP1 | |
2700092A01Rik; NAG20; nasopharyngeal carcinoma-associated gene 20 protein; UAP; Ubap; Ubap1; UBAP-1; ubiquitin associated protein 1; ubiquitin-associated protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51271 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.7 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NZ09 | |
UBAP1 | |
Recombinant protein corresponding to Human UBAP1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only