missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UBAP1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16604245
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16604245 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16604245 Supplier Invitrogen™ Supplier No. PA599994

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total UBAP1.

Originally named because of its sequence similarity to the Saccharomyces cerevisiae NHP2 (non-histone protein 2), this protein appears to be a highly conserved nuclear protein that is a component of the [U4/U6.U5] tri-snRNP. It binds to the 5' stem-loop of U4 snRNA. Two transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBAP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene UBAP1
Gene Accession No. Q5XIS7, Q8BH48, Q9NZ09
Gene Alias 2700092A01Rik; NAG20; nasopharyngeal carcinoma-associated gene 20 protein; UAP; Ubap; Ubap1; UBAP-1; ubiquitin associated protein 1; ubiquitin-associated protein 1
Gene Symbols UBAP1
Host Species Rabbit
Immunogen A synthesized peptide derived from human UBAP1(Accession Q9NZ09), corresponding to amino acid residues F460-S502.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362502, 51271, 67123
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.