missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UBAP2 Polyclonal Antibody

Product Code. 17712914
Change view
Click to view available options
Quantity:
20 μL
200 μL
Unit Size:
200µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17712914 20 μL 20µL
17780894 200 μL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17712914 Supplier Invitrogen™ Supplier No. PA5121513

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

UBAP2 Polyclonal Antibody for Western Blot, IHC (P)

The protein encoded by this gene contains a UBA (ubiquitin associated) domain, which is characteristic of proteins that function in the ubiquitination pathway. This gene may show increased expression in the adrenal gland and lymphatic tissues. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Sep 2013].
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBAP2
Applications Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene UBAP2
Gene Accession No. Q5T6F2
Gene Alias 1190005K07Rik; AA408600; AD-012 protein; AU045235; KIAA1491; Lig1; mKIAA1491; mLig-1; protein lingerer homolog 1; ubap2; UBAP-2; ubiquitin associated protein 2; ubiquitin-associated protein 2
Gene Symbols UBAP2
Host Species Rabbit
Immunogen Recombinant protein Ubiquitin Associated Protein 2. The antigen corresponds to amino acid range 1-104 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55833
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.