missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBAP2L Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-751V
This item is not returnable.
View return policy
Description
UBAP2L Polyclonal antibody specifically detects UBAP2L in Human samples. It is validated for Western Blot
Specifications
| UBAP2L | |
| Polyclonal | |
| Western Blot | |
| FLJ42300, KIAA0144NICE-4, NICE4, Protein NICE-4, ubiquitin associated protein 2-like, ubiquitin-associated protein 2-like | |
| The immunogen recognized by this antibody maps to a region between residue 1025 and the C-terminus (residue 1087) of human Ubiquitin Associated Protein 2-Like using the numbering given in entry AAH03170.1 (GeneID 9898). | |
| 0.1 mL | |
| Cell Biology | |
| 9898 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction