missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBAP2L Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81871DL594
This item is not returnable.
View return policy
Description
UBAP2L Polyclonal antibody specifically detects UBAP2L in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| UBAP2L | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| DyLight 594 | |
| FLJ42300, KIAA0144NICE-4, NICE4, Protein NICE-4, ubiquitin associated protein 2-like, ubiquitin-associated protein 2-like | |
| UBAP2L antibody was raised against a 19 amino acid peptide from near the amino terminus of human UBAP2L. The immunogen is located within amino acids 200 - 250 of UBAP2L. Amino Acid Squence: DDNYGNSSGNTWNNTGHFE | |
| 0.1 mL | |
| Cell Biology | |
| 9898 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction