missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ UBAP2L Antibody [PE], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NB100-750PE
This item is not returnable.
View return policy
Description
UBAP2L Polyclonal antibody specifically detects UBAP2L in Human samples. It is validated for Immunoprecipitation,Western Blot,KnockDown
Specifications
| UBAP2L | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot, KnockDown | |
| PE | |
| FLJ42300, KIAA0144NICE-4, NICE4, Protein NICE-4, ubiquitin associated protein 2-like, ubiquitin-associated protein 2-like | |
| The immunogen recognized by this antibody maps to a region between residues 400 and 450 of human Ubiquitin Associated Protein 2-Like using the numbering given in entry AAH03170.1 (GeneID 9898). | |
| 0.1 mL | |
| Cell Biology | |
| 9898 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction