missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UBASH3A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111061
This item is not returnable.
View return policy
Description
Immunogen sequence: LYSRDMRFVH YQTLRALFQY KPQNVDELTL SPGDYIFVDP TQQDEASEGW VIGISQRTGC RGFLPENYTD RASESDTWVK HRMY Highest antigen sequence indentity to the following orthologs: Rat - 88%, Mouse - 87%.
TULA (T cell ubiquitin ligand, UBASH3A, ubiquitin associated and SH3 domain containing A), STS-2 or CLIP4, is a 661 amino acid protein that localizes to both the nucleus and the cytoplasm and contains one SH3 domain and one UBA domain. Expressed at high levels in thymus, bone marrow, spleen and peripheral blood leukocytes, TULA exists as either a homodimer or a homo-oligomer that interferes with the degradation of receptor-type tyrosine kinases and promotes the accumulation of activated receptors on the cell surface. Additionally, TULA is part of an EGFR- and Cbl-containing complex that interacts with ubiquitinated proteins. The gene encoding TULA, which maps to human chromosome 21, may be involved in the pathogenesis of type I diabetes. Multiple isoforms of TULA are produced due to alternative splicing events.
Specifications
| UBASH3A | |
| Polyclonal | |
| Unconjugated | |
| UBASH3A | |
| 5830413C03Rik; C330001M22; Cbl-interacting protein 4; CLIP4; STS2; Sts-2; Suppressor of T-cell receptor signaling 2; T-cell ubiquitin ligand 1; T-cell ubiquitin ligand protein; TULA; TULA1; TULA-1; Ubash3a; ubiquitin associated and SH3 domain containing A; ubiquitin associated and SH3 domain containing, A; ubiquitin-associated and SH3 domain-containing protein A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 53347 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P57075 | |
| UBASH3A | |
| Recombinant Protein corresonding to Human UBASH3A. Recombinant protein control fragment (Product #RP-95890). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction