missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UbcH5a/UBE2D1 Antibody (OTI1E7), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74731AF405
This item is not returnable.
View return policy
Description
UbcH5a/UBE2D1 Monoclonal specifically detects UbcH5a/UBE2D1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| UbcH5a/UBE2D1 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| UBE2D1 | |
| Full length recombinant protein of human UbcH5a/UBE2D1 (NP_003329) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| OTI1E7 | |
| Western Blot | |
| E2(17)KB1, Stimulator of Fe transportUBC4/5 homolog, UBC4/5, UBC5A, UbcH5, UbcH5A, UBCH5AEC 6.3.2.19, UBCH5SFT, Ubiquitin carrier protein D1, ubiquitin-conjugating enzyme E2 D1, Ubiquitin-conjugating enzyme E2(17)KB 1, Ubiquitin-conjugating enzyme E2-17 kDa 1, ubiquitin-conjugating enzyme E2D 1 (UBC4/5 homolog, yeast), Ubiquitin-protein ligase D1 | |
| Mouse | |
| Affinity Purified | |
| Cancer, Hypoxia | |
| 7321.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction