missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2B Monoclonal Antibody (OTI1E11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803573S
This item is not returnable.
View return policy
Description
UBE2B Monoclonal antibody specifically detects UBE2B in Human samples. It is validated for Western BlotSpecifications
| UBE2B | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8WXC3 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 260434 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI1E11 | |
| Unconjugated | |
| PYDC1 | |
| 2610301N02Rik; E2 protein; E2 ubiquitin-conjugating enzyme B; E2(14k); E214K; E2-14k; E2-17kDa; HHR6B; HR6B; mHR6B; RAD6 homolog B; Rad6b; UBC2; Ube2b; Ubiquitin carrier protein B; ubiquitin conjugating enzyme; ubiquitin conjugating enzyme E2 B; ubiquitin conjugating enzyme E2B; ubiquitin-conjugating enzyme E2 B; Ubiquitin-conjugating enzyme E2(14k); ubiquitin-conjugating enzyme E2-17 kDa; ubiquitin-conjugating enzyme E2B; ubiquitin-conjugating enzyme E2B (RAD6 homolog); ubiquitin-conjugating enzyme E2B (RAD6 homolog, S. cerevisiae); ubiquitin-conjugating enzyme E2B, RAD6 homolog; ubiquitin-conjugating enzyme E2B, RAD6 homology; ubiquitin-protein ligase B | |
| Full length human recombinant protein of human PYDC1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction