Learn More
ZRANB1 Monoclonal Antibody (OTI2A10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA810024
Encompass_Preferred
 View more versions of this product
Product Code. 17592754
  • € 540.87 / 100µL
Description
ZRANB1 is a positive regulator of the Wnt signaling pathway that specifically cleaves ′Lys-63′-linked ubiquitin chains. It acts by deubiquitinating APC protein, a negative regulator of Wnt-mediated transcription and may also modulate TNF-alpha signaling.
Specifications
Specifications
ZRANB1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9UGI0
ZRANB1
Human recombit protein fragment corresponding to amino acids 208-296 of human ZRANB1 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI2A10
Unconjugated
ZRANB1
TRABID
Mouse
Affinity Chromatography
RUO
54764
-20° C, Avoid Freeze/Thaw Cycles