missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UBE2C Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142493
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize isoforms 1 (NP_008950.1), 2 ( NP_861515.1) and 4 (NP_861517.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:8,000.
This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. The encoded protein is required for the destruction of mitotic cyclins and for cell cycle progression, and may be involved in cancer progression. Accepts ubiquitin from the E1 complex and catalyzes its covalent attachment to other proteins. In vitro catalyzes 'Lys-11'- and 'Lys-48'-linked polyubiquitination. Acts as an essential factor of the anaphase promoting complex/cyclosome (APC/C), a cell cycle-regulated ubiquitin ligase that controls progression through mitosis. Acts by initiating 'Lys-11'-linked polyubiquitin chains on APC/C substrates, leading to the degradation of APC/C substrates by the proteasome and promoting mitotic exit.
Specifications
| UBE2C | |
| Polyclonal | |
| Unconjugated | |
| UBE2C | |
| (E3-independent) E2 ubiquitin-conjugating enzyme C; 1110015A16Rik; cyclin-selective ubiquitin carrier protein; D2Ertd695e; dJ447F3.2; E2 ubiquitin-conjugating enzyme C; mitotic-specific ubiquitin-conjugating enzyme; RP3-447F3.1; ubcH10; UBE2C; Ubiquitin carrier protein C; ubiquitin conjugating enzyme E2 C; ubiquitin conjugating enzyme E2C; ubiquitin-conjugating enzyme E2 C; ubiquitin-conjugating enzyme E2C; ubiquitin-protein ligase C | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 11065 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O00762 | |
| UBE2C | |
| Peptide with sequence C-SGDKGISAFPESDN. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction