missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPIL1 Antibody (003), DyLight 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90270V
This item is not returnable.
View return policy
Description
PPIL1 Monoclonal specifically detects PPIL1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| PPIL1 | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 51645 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 3 | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CYPL1cyclophilin-related gene 1, EC 5.2.1.8, hCyPX, MGC678, peptidyl-prolyl cis-trans isomerase, peptidyl-prolyl cis-trans isomerase-like 1, peptidylprolyl isomerase (cyclophilin)-like 1, PPIase, Rotamase PPIL1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human PPIL1 (Q9Y3C6; Met1-Gly166). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction