missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2CBP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 292.00 - € 572.00
Specifications
| Antigen | UBE2CBP |
|---|---|
| Dilution | Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18434711
|
Novus Biologicals
NBP1-86757-25ul |
25 μL |
€ 292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18280907
|
Novus Biologicals
NBP1-86757 |
0.1 mL |
€ 572.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UBE2CBP Polyclonal specifically detects UBE2CBP in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| UBE2CBP | |
| Polyclonal | |
| Rabbit | |
| Human | |
| C6orf157, chromosome 6 open reading frame 157, EC 6.3.2, EC 6.3.2.-, H10BHDKFZp434A1520, MGC126561, MGC126587, UBCH10 binding protein with a hect-like domain, UbcH10-binding protein with a HECT-like domain, ubiquitin-conjugating enzyme E2C binding protein, ubiquitin-conjugating enzyme E2C-binding protein, YJR141W | |
| UBE3D | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 90025 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:SLVIESLRNSKYIKKFPLLENTFKADSSSAWSAVKVLYQPCIKSRNEKLVSLWESDISVHPLTLPSATCLELLL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title