missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UBE2D1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596006
This item is not returnable.
View return policy
Description
Immunogen sequence: MALKRIQKEL SDLQRDPPAH CSAGPVGDDL FHWQATIMGP PDSAYQGGVF FLTVHFPTDY PFKPPKIAFT TKIYHPNINS NGSICLDILR SQWSPALTVS KVLLSICSLL CDPNPDDPLV PDIAQIYKSD KEKYNRHARE WTQKYAM; Positive Samples: SW620, BT-474; Cellular Location: Cytoplasm.
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is closely related to a stimulator of iron transport (SFT), and is up-regulated in hereditary hemochromatosis. It also functions in the ubiquitination of the tumor-suppressor protein p53 and the hypoxia-inducible transcription factor HIF1alpha by interacting with the E1 ubiquitin-activating enzyme and the E3 ubiquitin-protein ligases.
Specifications
| UBE2D1 | |
| Polyclonal | |
| Unconjugated | |
| UBE2D1 | |
| (E3-independent) E2 ubiquitin-conjugating enzyme D1; E2 ubiquitin-conjugating enzyme D1; E2(17)KB 1; E2(17)KB1; SFT; Stimulator of Fe transport; UBC 4/5; UBC4/5; UBC4/5 homolog; UBC5A; UBCH 5; UBCH 5A; UBCH5; UBCH5A; UBE2D1; ubiquitin; Ubiquitin carrier protein D1; ubiquitin conjugating enzyme E2 D1; ubiquitin conjugating enzyme E2D 1; ubiquitin-conjugating enzyme E2 D1; Ubiquitin-conjugating enzyme E2(17)KB 1; ubiquitin-conjugating enzyme E2-17 kDa 1; ubiquitin-conjugating enzyme E2D 1; ubiquitin-conjugating enzyme E2D 1 (UBC4/5 homolog, yeast); ubiquitin-conjugating enzyme E2D 1, UBC4/5 homolog; ubiquitin-protein ligase D1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 216080, 361831, 7321 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| D3ZDK2, P51668, P61080 | |
| UBE2D1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-147 of human UBE2D1 (NP_0033291). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction