missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UBE2D2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106767
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total UBE2D2.
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. This enzyme functions in the ubiquitination of the tumor-suppressor protein p53, which is induced by an E3 ubiquitin-protein ligase. Two alternatively spliced transcript variants have been found for this gene and they encode distinct isoforms.
Specifications
| UBE2D2 | |
| Polyclonal | |
| Unconjugated | |
| UBE2D2 | |
| (E3-independent) E2 ubiquitin-conjugating enzyme D2; 1500034D03Rik; E2 ubiquitin conjugating enzyme UbcH5B; E2 ubiquitin-conjugating enzyme D2; E2(17)KB 2; E2(17)KB2; E217kB; p53-regulated ubiquitin-conjugating enzyme 1; PUBC1; similar to ubiquitin-conjugating enzyme E2D 2; Ubc2e; ubc2e ubiquitin conjugating enzyme; ubc4; UBC4/5; UBC5B; UBCH4; UBCH5B; ube2d2; Ube2d2a; Ubiquitin carrier protein D2; ubiquitin conjugating enzyme 2e; ubiquitin conjugating enzyme E2 D2; ubiquitin conjugating enzyme E2D; ubiquitin conjugating enzyme E2D 2; ubiquitin-conjugating enzyme E2 D2; Ubiquitin-conjugating enzyme E2(17)KB 2; ubiquitin-conjugating enzyme E2-17 kDa 2; ubiquitin-conjugating enzyme E2D 2; ubiquitin-conjugating enzyme E2D 2 (homologous to yeast UBC4/5); ubiquitin-conjugating enzyme E2D 2 (UBC4/5 homolog, yeast); ubiquitin-conjugating enzyme E2D 2A; ubiquitin-protein ligase D2; Unknown (protein for MGC:138112); zgc:55886; zgc:55886 protein; zgc:77149; zmp:0000000618 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 56550, 641452, 7322 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P62837, P62838, P62839 | |
| UBE2D2, Ube2d2a | |
| A synthesized peptide derived from human UBE2D2(Accession P62837), corresponding to amino acid residues I106-M147. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction