Learn More
Invitrogen™ UBE2D4 Recombinant Rabbit Monoclonal Antibody (129)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA530982
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human UBE2D4.
Accepts ubiquitin from the E1 complex and catalyzes its covalent attachment to other proteins. In vitro able to promote polyubiquitination using all 7 ubiquitin Lys residues, but may prefer 'Lys-11' and 'Lys-48'-linked polyubiquitination.
Specifications
| UBE2D4 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q9Y2X8 | |
| Ube2d4 | |
| Recombinant Human UBE2D4 protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA | |
| 129 | |
| Unconjugated | |
| Ube2d4 | |
| E2 ubiquitin-conjugating enzyme D2B; E2 ubiquitin-conjugating enzyme D4; E2(17)KB 2B; HBUCE1; RGD69425; UBCH5D; Ube2d2; Ube2d2b; UBE2D4; Ubiquitin carrier protein D2B; ubiquitin carrier protein D4; ubiquitin conjugating enzyme E2 D4 (putative); ubiquitin conjugating enzyme E2D 4 (putative); ubiquitin-conjugating enzyme E2 D2B; Ubiquitin-conjugating enzyme E2 D4; ubiquitin-conjugating enzyme E2(17)KB 2B; Ubiquitin-conjugating enzyme E2-17 kDa 2B; ubiquitin-conjugating enzyme E2D 2; ubiquitin-conjugating enzyme E2D 4 (putative); ubiquitin-conjugating enzyme HBUCE1; ubiquitin-protein ligase D2B; Ubiquitin-protein ligase D4 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 51619 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.