missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2E3 Monoclonal Antibody (OTI1D10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504679S
This item is not returnable.
View return policy
Description
UBE2E3 Monoclonal antibody specifically detects UBE2E3 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| UBE2E3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P48052 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1358 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI1D10 | |
| Unconjugated | |
| CPA2 | |
| E2 ubiquitin-conjugating enzyme E3; Ubce4; UBCH9; UbcM2; UBE2E3; Ubiquitin carrier protein E3; ubiquitin conjugating enzyme E2 E3; ubiquitin conjugating enzyme E2E 3; ubiquitin-conjugating enzyme 4; ubiquitin-conjugating enzyme E2 E3; Ubiquitin-conjugating enzyme E2-23 kDa; ubiquitin-conjugating enzyme E2E 3; ubiquitin-conjugating enzyme E2E 3 (homologous to yeast UBC4/5); ubiquitin-conjugating enzyme E2E 3 (UBC4/5 homolog, yeast); ubiquitin-conjugating enzyme E2E 3, UBC4/5 homolog; ubiquitin-protein ligase E3 | |
| Full length human recombinant protein of human CPA2 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction