missing translation for 'onlineSavingsMsg'
Learn More

UBE2E3 Monoclonal Antibody (OTI6D3), TrueMAB™, OriGene

Product Code. p-200015583
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17577502 30 μL 30µL
17557502 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17577502 Supplier Origene Technologies Supplier No. TA800071S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

UBE2E3 Monoclonal antibody specifically detects UBE2E3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen UBE2E3
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI6D3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Uqcrc1
Gene Accession No. P31930
Gene Alias E2 ubiquitin-conjugating enzyme E3; Ubce4; UBCH9; UbcM2; UBE2E3; Ubiquitin carrier protein E3; ubiquitin conjugating enzyme E2 E3; ubiquitin conjugating enzyme E2E 3; ubiquitin-conjugating enzyme 4; ubiquitin-conjugating enzyme E2 E3; Ubiquitin-conjugating enzyme E2-23 kDa; ubiquitin-conjugating enzyme E2E 3; ubiquitin-conjugating enzyme E2E 3 (homologous to yeast UBC4/5); ubiquitin-conjugating enzyme E2E 3 (UBC4/5 homolog, yeast); ubiquitin-conjugating enzyme E2E 3, UBC4/5 homolog; ubiquitin-protein ligase E3
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 46-305 of human UQCRC1 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7384
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.