missing translation for 'onlineSavingsMsg'
Learn More

UBE2G2 Antibody [Janelia Fluor« 669], Novus Biologicals Biologicals™

Product Code. 30488931 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30488931 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30488931 Supplier Novus Biologicals Supplier No. NBP335140JF669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

UBE2G2 Polyclonal antibody specifically detects UBE2G2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2G2
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias EC 6.3.2.19, UBC7human ubiquitin conjugating enzyme G210ubiquitin-conjugating enzyme E2G 2 (homologous to yeast UBC7), Ubiquitin carrier protein G2, ubiquitin conjugating enzyme 7, ubiquitin conjugating enzyme G2, ubiquitin-conjugating enzyme E2 G2, ubiquitin-conjugating enzyme E2G 2 (UBC7 homolog, yeast), Ubiquitin-protein ligase G2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-165 of human UBE2G2 (NP_003334.2).,, Sequence:, MAGTALKRLMAEYKQLTLNPPEGIVAGPMNEENFFEWEALIMGPEDTCFEFGVFPAILSFPLDYPLSPPKMRFTCEMFHPNIYPDGRVCISILHAPGDDPMGYESSAERWSPVQSVEKILLSVVSMLAEPNDESGANVDASKMWRDDREQFYKIAKQIVQKSLGL
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 7327
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.