missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2I/Ubc9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 353.00 - € 480.00
Specifications
| Antigen | UBE2I/Ubc9 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18491181
|
Novus Biologicals
NBP1-86887-25ul |
25 μL |
€ 353.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18287098
|
Novus Biologicals
NBP1-86887 |
0.1 mL |
€ 480.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UBE2I/Ubc9 Polyclonal specifically detects UBE2I/Ubc9 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| UBE2I/Ubc9 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| C358B7.1, EC 6.3.2, EC 6.3.2.-, P18, SUMO-1-protein ligase, SUMO-protein ligase, UBC9SUMO-conjugating enzyme UBC9, UBCE9, Ubiquitin carrier protein 9, Ubiquitin carrier protein I, ubiquitin conjugating enzyme 9, Ubiquitin-conjugating enzyme E2 I, ubiquitin-conjugating enzyme E2I (homologous to yeast UBC9), ubiquitin-conjugating enzyme E2I (UBC9 homolog, yeast), ubiquitin-conjugating enzyme UbcE2A, ubiquitin-like protein SUMO-1 conjugating enzyme, ubiquitin-protein ligase E2I, Ubiquitin-protein ligase I | |
| UBE2I | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Rabbit | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 7329 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:IALSRLAQERKAWRKDHPFGFVAVPTKNPDGTMNLMNWECAIPGKKGTPWEGGLFKLRMLFKDDYPSSPPKCKFEPPLFHPNVYPSGTVCLSILEEDKDWRPAITIKQILLGIQELLNEPNIQDPAQAEAYTIYCQNRVEYEKRVRAQ | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title