missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2I/Ubc9 Antibody (SAIC-36A-9), Janelia Fluor™ 646 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20104JF646
This item is not returnable.
View return policy
Description
UBE2I/Ubc9 Monoclonal antibody specifically detects UBE2I/Ubc9 in Human samples. It is validated for Western Blot, ELISA
Specifications
| UBE2I/Ubc9 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| C358B7.1, EC 6.3.2, EC 6.3.2.-, P18, SUMO-1-protein ligase, SUMO-protein ligase, UBC9SUMO-conjugating enzyme UBC9, UBCE9, Ubiquitin carrier protein 9, Ubiquitin carrier protein I, ubiquitin conjugating enzyme 9, Ubiquitin-conjugating enzyme E2 I, ubiquitin-conjugating enzyme E2I (homologous to yeast UBC9), ubiquitin-conjugating enzyme E2I (UBC9 homolog, yeast), ubiquitin-conjugating enzyme UbcE2A, ubiquitin-like protein SUMO-1 conjugating enzyme, ubiquitin-protein ligase E2I, Ubiquitin-protein ligase I | |
| Peptide "e;GLFIIDGK"e;, derived from the SUMO-conjugating enzyme UBC9, conjugated to KLH. | |
| 0.1 mL | |
| Biologically Active Proteins | |
| 7329 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Western Blot, ELISA | |
| SAIC-36A-9 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction