Learn More
UBE2J2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17713-1-AP-20UL
 View more versions of this product
Product Code. 16837654
  • € 145.00 / 20µL
Description
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. UBE2J2 is a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is located in the membrane of the endoplasmic reticulum.The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is located in the membrane of the endoplasmic reticulum. Multiple alternatively spliced transcript variants have been found for this gene, but the full-length nature of some variants has not been defined.
Specifications
Specifications
UBE2J2
Polyclonal
Unconjugated
UBE2J2
NCUBE 2, NCUBE2, PRO2121, UBE2J2
Rabbit
Antigen Affinity Chromatography
RUO
118424, 140499, 298689
-20°C
Liquid
Western Blot
0.41 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6P073, Q8N2K1
Ube2j2
UBE2J2 Fusion Protein Ag12039
20 μL
Primary
Human, Rat, Mouse