missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2K/E2-25K Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB300-840
This item is not returnable.
View return policy
Description
UBE2K/E2-25K Polyclonal specifically detects UBE2K/E2-25K in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| UBE2K/E2-25K | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| E2(25K), E2-25K, EC 6.3.2.19, HIP-2, huntingtin interacting protein 2, Huntingtin-interacting protein 2, HYPG, ubiquitin carrier protein, ubiquitin-conjugating enzyme E2 K, Ubiquitin-conjugating enzyme E2(25K), Ubiquitin-conjugating enzyme E2-25 kDa, Ubiquitin-conjugating enzyme E2-25K, ubiquitin-conjugating enzyme E2K (UBC1 homolog, yeast), ubiquitin-protein ligase | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3093 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
| 0.5 mg/mL | |
| Western Blot 1 - 3 ug/ml, Immunohistochemistry 1 - 2 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml, Peptide ELISA Detection limit 1:2000 | |
| NP_005330.1 | |
| UBE2K | |
| Peptide with sequence C-SWDVETATELLLSN corresponding to C-Terminus according to NP_005330.1, NP_001104582.1, NP_001104583.1. | |
| 0.1 mg | |
| Primary | |
| This antibody is expected to recognize isoform 1 (NP_005330.1), isoform 2 (NP_001104582.1) and isoform 3 (NP_001104583.1). | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction