missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2L3, Mouse anti-Human, Clone: 3B7, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00007332-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: MAASRRLMKELEEIRKCGMKNFRNIQVDEANLLTWQGLIVPDNPPYDKGAFRIEINFPAEYPFKPPKITFKTKIYHPNIDEKGQVCLPVISAENWKPATKTDQVIQSLIALVNDPQPEHPLRADLAEEYSKDRKKFCKNAEEFTKKYGEKRPVDSpecifications
| UBE2L3 | |
| Monoclonal | |
| Unconjugated | |
| ubiquitin-conjugating enzyme E2L 3 | |
| E2-F1/L-UBC/UBCH7/UbcM4 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 3B7 | |
| In 1x PBS, pH 7.4 | |
| BC053368 | |
| UBE2L3 | |
| UBE2L3 (AAH53368, 1 a.a. ∼ 154 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 7332 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction