Learn More
Invitrogen™ UBE2L3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530202
Description
Recommended positive controls: 293T, HeLa, JurKat, mouse testis, UBE2L3 shRNA-transfected 293T. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (99%), Pig (100%), Chicken (100%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes (E1s), ubiquitin-conjugating enzymes (E2s) and ubiquitin-protein ligases (E3s). UBE2L3 is a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is demonstrated to participate in the ubiquitination of p53, c-Fos, and the NF-kB precursor p105 in vitro.
Specifications
| UBE2L3 | |
| Polyclonal | |
| Unconjugated | |
| UBE2L3 | |
| C79827; E2 ubiquitin-conjugating enzyme L3; E2-F1; L-UBC; UBCE7; UBCH7; UbcM4; Ube2l3; Ubiquitin carrier protein L3; ubiquitin conjugating enzyme E2 L3; ubiquitin conjugating enzyme E2L 3; ubiquitin-conjugating enzyme 7; ubiquitin-conjugating enzyme E2 L3; Ubiquitin-conjugating enzyme E2-F1; ubiquitin-conjugating enzyme E2L 3; ubiquitin-conjugating enzyme UBCH7; ubiquitin-protein ligase L3; Unknown (protein for MGC:127956) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22195, 7332 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.78 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| P68036, P68037 | |
| UBE2L3 | |
| Recombinant protein encompassing a sequence within the center region of human UBE2L3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.