missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2L3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14415-1-AP-150UL
This item is not returnable.
View return policy
Description
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes (E1s), ubiquitin-conjugating enzymes (E2s) and ubiquitin-protein ligases (E3s). This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is demonstrated to participate in the ubiquitination of p53, c-Fos, and the NF-kB precursor p105 in vitro. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.Specifications
| UBE2L3 | |
| Polyclonal | |
| Unconjugated | |
| UBE2L3 | |
| E2 F1, L UBC, UBCE7, UBCH7, UbcM4, UBE2L3, Ubiquitin carrier protein L3, Ubiquitin protein ligase L3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22195, 363836, 7332 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P68036, P68037 | |
| Ube2l3 | |
| UBE2L3 Fusion Protein Ag5758 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction