missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2M Mouse anti-Canine, Human, Mouse, Clone: OTI2D9, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503770
This item is not returnable.
View return policy
Description
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. The encoded protein is linked with a ubiquitin-like protein, NEDD8, which can be conjugated to cellular proteins, such as Cdc53/culin.Specifications
| UBE2M | |
| Monoclonal | |
| 0.69 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P61081, P61082 | |
| UBE2M | |
| Full length human recombit protein of human UBE2M produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Canine, Human, Mouse | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI2D9 | |
| Unconjugated | |
| UBE2M | |
| UBC-RS2, UBC12, hUbc12 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 22192, 484221, 9040 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction