missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2M/Ubc12 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97606FR
This item is not returnable.
View return policy
Description
UBE2M/Ubc12 Polyclonal specifically detects UBE2M/Ubc12 in Human samples. It is validated for Western Blot.
Specifications
| UBE2M/Ubc12 | |
| Polyclonal | |
| Western Blot | |
| EC 6.3.2, EC 6.3.2.-, hUbc12, NEDD8 carrier protein, NEDD8 protein ligase, UBC12NEDD8-conjugating enzyme Ubc12, UBC-RS2, ubiquitin carrier protein M, Ubiquitin-conjugating enzyme E2 M, ubiquitin-conjugating enzyme E2M (homologous to yeast UBC12), ubiquitin-conjugating enzyme E2M (UBC12 homolog, yeast), ubiquitin-protein ligase M, yeast UBC12 homolog | |
| Produced in rabbits immunized with E. coli-derived Human UBE2M/Ubc12 fragment. | |
| 0.1 mL | |
| Biologically Active Proteins | |
| 9040 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction