missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2N/Ubc13 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98828AF750
This item is not returnable.
View return policy
Description
UBE2N/Ubc13 Polyclonal antibody specifically detects UBE2N/Ubc13 in Human samples. It is validated for Western Blot
Specifications
| UBE2N/Ubc13 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 750 | |
| bendless-like ubiquitin conjugating enzyme, Bendless-like ubiquitin-conjugating enzyme, BLU, EC 6.3.2.19, MGC131857, MGC8489, UBC13, UbcH-ben, Ubiquitin carrier protein N, ubiquitin-conjugating enzyme E2 N, ubiquitin-conjugating enzyme E2N (homologous to yeast UBC13), ubiquitin-conjugating enzyme E2N (UBC13 homolog, yeast), Ubiquitin-protein ligase N, yeast UBC13 homolog | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human UBE2N/Ubc13. | |
| 0.1 mL | |
| Cancer | |
| 7334 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction