Learn More
UBE2N/Ubc13 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-98828PCP
Product Code. 18630576
  • € 407.00 / 0.10mL
Description
UBE2N/Ubc13 Polyclonal antibody specifically detects UBE2N/Ubc13 in Human samples. It is validated for Western Blot