missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UBE2Q1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15809013
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15809013 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15809013 Supplier Invitrogen™ Supplier No. PA570600

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 100%; Rat: 93%; Yeast: 100%.

The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes (E1s), ubiquitin-conjugating enzymes (E2s), and ubiquitin-protein ligases (E3s). This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. The encoded protein is 98% identical to the mouse counterpart.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2Q1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene UBE2Q1
Gene Accession No. Q7Z7E8
Gene Alias 1110002C01Rik; 2310012M18Rik; AI854014; E2 ubiquitin-conjugating enzyme Q1; galactosyl transferase-associated protein; GTAP; hypothetical protein LOC553603; LOW QUALITY PROTEIN: ubiquitin-conjugating enzyme E2 Q1; NICE5; NICE-5; PRO3094; protein NICE-5; UBE2Q; UBE2Q1; Ubiquitin carrier protein Q1; ubiquitin conjugating enzyme E2 Q1; ubiquitin conjugating enzyme E2Q family member 1; ubiquitin-conjugating enzyme E2 Q1; ubiquitin-conjugating enzyme E2 Q1; ubiquitin-conjugating enzyme E2 Q2; ubiquitin-conjugating enzyme E2Q (putative) 1; ubiquitin-conjugating enzyme E2Q family member 1; ubiquitin-protein ligase Q1; wu:fi42b06; zgc:110254
Gene Symbols UBE2Q1
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human UBE2Q1 (aa 174-223).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55585
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.